3-Bromo-5-(trifluoromethyl)-1H-indazole

Supplier: Apollo Scientific
Warning

PC3261-1G PC3261-250MG
APOSPC3261-1GEA 92.8 EUR
APOSPC3261-1G APOSPC3261-250MG
3-Bromo-5-(trifluoromethyl)-1H-indazole
3-Bromo-5-(trifluoromethyl)-1H-indazole
Formula: C₈H₄BrF₃N₂
MW: 265.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998163
CAS Number: 1086378-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR