Ethyl trifluoroacetate 99%

Supplier: Apollo Scientific
Danger

PC3270-100G PC3270-1KG PC3270-500G
APOSPC3270-100GEA 21.9 EUR
APOSPC3270-100G APOSPC3270-1KG APOSPC3270-500G
Ethyl trifluoroacetate 99%
Ethyl trifluoroacetate
Formula: C₄H₅F₃O₂
Boiling Pt: 60…62 °C
Melting Pt: –78 °C
Density: 1,194 g/cm³ (25 °C)
Flash Pt: –1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000419
CAS Number: 383-63-1
EINECS: 206-851-6
UN: 2924
ADR: 3+8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR