Ethyl-5-(trifluoromethoxy)indole-2-carboxylate

Supplier: Apollo Scientific
Warning

PC3285E-1G PC3285E-5G
APOSPC3285E-1GEA 116 EUR
APOSPC3285E-1G APOSPC3285E-5G
Ethyl-5-(trifluoromethoxy)indole-2-carboxylate
Ethyl-5-(trifluoromethoxy)indole-2-carboxylate
Formula: C₁₂H₁₀F₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00219837
CAS Number: 175203-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR