Ethyl-4-(trifluoromethyl)benzoate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 4-(trifluoromethyl)benzoate
PC3288-1G
PC3288-5G
APOSPC3288-1GEA
14.7
EUR
APOSPC3288-1G
APOSPC3288-5G
Ethyl-4-(trifluoromethyl)benzoate 97%
Ethyl 4-(trifluoromethyl)benzoate
Formula:
C₁₀H₉F₃O₂ MW: 218,18 g/mol Boiling Pt: 78…80 °C/5mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00013563 CAS Number: 583-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...