Iron(III) fluoride, anhydrous 99%

Supplier: Apollo Scientific
Danger

Synonyms: Iron trifluoride, Ferric fluoride

PC3291-100G PC3291-50G
APOSPC3291-100GEA 219 EUR
APOSPC3291-100G APOSPC3291-50G
Iron(III) fluoride, anhydrous 99%
Iron (III) fluoride
Formula: F₃Fe
Melting Pt: 1030 °C
Density: 3,52 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011012
CAS Number: 7783-50-8
EINECS: 232-002-4
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR