4-Fluoroanisole 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Fluoro-4-methoxybenzene
PC3440-1KG
PC3440-250G
PC3440-500G
APOSPC3440-1KGEA
123
EUR
APOSPC3440-1KG
APOSPC3440-250G
APOSPC3440-500G
4-Fluoroanisole 99%
4-Fluoroanisole
Formula:
C₇H₇FO MW: 126,13 g/mol Boiling Pt: 156…158 °C Melting Pt: –45 °C Density: 1,116 g/cm³ (25 °C) Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000348 CAS Number: 459-60-9 UN: 3271 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...