3-Fluorobenzal chloride 98%

Supplier: Apollo Scientific

Synonyms: α,α-Dichloro-3-fluortoluene, 1-(Dichloromethyl)-3-fluorobenzene

PC3444B-25G PC3444B-5G
APOSPC3444B-25GEA 263 EUR
APOSPC3444B-25G APOSPC3444B-5G
3-Fluorobenzal chloride 98%
3-Fluorobenzal chloride
Formula: C₇H₅Cl₂F
MW: 179,02 g/mol
Boiling Pt: 195…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00037112
CAS Number: 402-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR