1-(4-Fluoro-2-nitrophenyl)piperidine

Supplier: Apollo Scientific
Warning

PC3451-1G PC3451-25G PC3451-5G
APOSPC3451-1GEA 65.5 EUR
APOSPC3451-1G APOSPC3451-25G APOSPC3451-5G
1-(4-Fluoro-2-nitrophenyl)piperidine
1-(4-Fluoro-2-nitrophenyl)piperidine
Formula: C₁₁H₁₃FN₂O₂
MW: 224,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00798370
CAS Number: 719-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR