4-Fluorobenzaldehyde 99%
Supplier: Apollo Scientific
|
Warning
|
PC3470-100G
PC3470-500G
APOSPC3470-100GEA
21.9
EUR
APOSPC3470-100G
APOSPC3470-500G
4-Fluorobenzaldehyde 99%
4-Fluorobenzaldehyde
Synthesis, 63 (1991): simple preparation of several 4-aryloxy phenols (via substitution then Baeyer-Villiger).
Formula:
C₇H₅FO MW: 124,11 g/mol Boiling Pt: 70…72 °C/10mmHg Melting Pt: –10 °C Density: 1,175 g/cm³ (25 °C) Flash Pt: 56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003378 CAS Number: 459-57-4 EINECS: 207-293-6 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...