N-Fluorobenzenesulphonimide 98%
Supplier: Apollo Scientific
|
Warning
|
PC3487-25G
PC3487-100G
PC3487-500G
PC3487-1KG
APOSPC3487-25GEA
20.5
EUR
APOSPC3487-25G
APOSPC3487-100G
APOSPC3487-500G
APOSPC3487-1KG
N-Fluorobenzenesulphonimide 98%
N-Fluorobenzenesulphonimide
Electrophilic fluorinating agent. Ask for tech. literature .60.2mg/g active fluorine. Solubility: THF 0.24g/ml; CH2Cl2 0.4g/ml; CH3CN 0.25g/ml; Toluene 0.09g/ml.
Formula:
C₁₂H₁₀FNO₄S₂ MW: 315,34 g/mol Melting Pt: 110…118 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00144885 CAS Number: 133745-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...