2-Fluorobenzhydrazide 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluorobenzohydrazide, 2-Fluorobenzene-1-carbohydrazide, 2-Fluorobenzoic hydrazide, 2-Fluorobenzoic acid hydrazide

PC3495E-10G PC3495E-50G PC3495E-1G
APOSPC3495E-10GEA 72.7 EUR
APOSPC3495E-10G APOSPC3495E-50G APOSPC3495E-1G
2-Fluorobenzhydrazide 98%
2-Fluorobenzhydrazide
Formula: C₇H₇FN₂O
MW: 154,14 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00025112
CAS Number: 446-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR