1-(2-Fluoro-4-nitrophenyl)homopiperazine
Supplier: Apollo Scientific
PC3522-1G
PC3522-5G
APOSPC3522-1GEA
137
EUR
APOSPC3522-1G
APOSPC3522-5G
1-(2-Fluoro-4-nitrophenyl)homopiperazine
1-(2-Fluoro-4-nitrophenyl)homopiperazine
Formula:
C₁₁H₁₄FN₃O₂ Melting Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD08692379 CAS Number: 392660-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...