2-Fluorobenzonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
o-Fluorobenzonitrile
PC3530-100G
PC3530-500G
APOSPC3530-100GEA
26.1
EUR
APOSPC3530-100G
APOSPC3530-500G
2-Fluorobenzonitrile 98%
2-Fluorobenzonitrile
Formula:
C₇H₄FN MW: 121,11 g/mol Boiling Pt: 92…95 °C/22mmHg Density: 1,138 g/cm³ (25 °C) Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001773 CAS Number: 394-47-8 EINECS: 206-897-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...