2'-Chloro-5'-fluoroacetophenone 99%

Supplier: Apollo Scientific
Warning

PC3531-25G PC3531-5G PC3531-100G
APOSPC3531-25GEA 280 EUR
APOSPC3531-25G APOSPC3531-5G APOSPC3531-100G
2'-Chloro-5'-fluoroacetophenone 99%
2'-Chloro-5'-fluoroacetophenone
Formula: C₈H₆ClFO
MW: 172,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042574
CAS Number: 2965-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR