3-Fluorobenzonitrile 98%

Supplier: Apollo Scientific
Danger

PC3540-100G PC3540-25G PC3540-500G
APOSPC3540-100GEA 52.2 EUR
APOSPC3540-100G APOSPC3540-25G APOSPC3540-500G
3-Fluorobenzonitrile 98%
3-Fluorobenzonitrile
Formula: C₇H₄FN
MW: 121,11 g/mol
Boiling Pt: 182…183 °C
Melting Pt: 15…16 °C
Density: 1,134 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00001797
CAS Number: 403-54-3
EINECS: 206-963-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR