5-Fluoro-3-pyridineboronic acid 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Fluoropyridine-3-boronic acid, (5-Fluoropyridin-3-yl)boronic acid

PC3624-1G PC3624-5G PC3624-25G
APOSPC3624-1GEA 29 EUR
APOSPC3624-1G APOSPC3624-5G APOSPC3624-25G
5-Fluoro-3-pyridineboronic acid 95+%
5-Fluoro-3-pyridineboronic acid
Formula: C₅H₅BFNO₂
Melting Pt: 233…236 °C
Storage Temperature: Ambient
MDL Number: MFCD07368243
CAS Number: 872041-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR