5-(Trifluoromethoxy)isatin 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-(Trifluoromethoxy)indoline-2,3-dione
PC3629-1G
PC3629-5G
APOSPC3629-1GEA
58.2
EUR
APOSPC3629-1G
APOSPC3629-5G
5-(Trifluoromethoxy)isatin 97%
5-(Trifluoromethoxy)isatin
Formula:
C₉H₄F₃NO₃ MW: 231,13 g/mol Melting Pt: 170…172 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00192524 CAS Number: 169037-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...