1-(4-Fluorobenzoyl)piperidine 98%

Supplier: Apollo Scientific
Warning

PC3633-5G PC3633-100G PC3633-25G
APOSPC3633-5GEA 68.3 EUR
APOSPC3633-5G APOSPC3633-100G APOSPC3633-25G
1-(4-Fluorobenzoyl)piperidine 98%
1-(4-Fluorobenzoyl)piperidine
Formula: C₁₂H₁₄BrNO
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00174322
CAS Number: 58547-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR