2-Fluorobenzyl alcohol 97%
Supplier: Apollo Scientific
|
Warning
|
PC3640-100G
PC3640-25G
APOSPC3640-100GEA
49.3
EUR
APOSPC3640-100G
APOSPC3640-25G
2-Fluorobenzyl alcohol 97%
2-Fluorobenzyl alcohol
Formula:
C₇H₇FO Boiling Pt: 97…101 °C/21mmHg Density: 1,175 g/cm³ (25 °C) Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004601 CAS Number: 446-51-5 EINECS: 207-170-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...