1-(5-Fluoro-2-methylphenyl)pyrrolidine

Supplier: Apollo Scientific

PC3654-1G PC3654-5G
APOSPC3654-1GEA 132 EUR
APOSPC3654-1G APOSPC3654-5G
1-(5-Fluoro-2-methylphenyl)pyrrolidine
1-(5-Fluoro-2-methylphenyl)pyrrolidine
Formula: C₁₁H₁₄FN
Storage Temperature: Ambient
MDL Number: MFCD09878397
CAS Number: 1000339-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR