3-Fluorobenzylamine 95%

Supplier: Apollo Scientific
Danger

PC3680-25G PC3680-5G PC3680-100G
APOSPC3680-25GEA 60.9 EUR
APOSPC3680-25G APOSPC3680-5G APOSPC3680-100G
3-Fluorobenzylamine 95%
3-Fluorobenzylamine
Formula: C₇H₈FN
Boiling Pt: 87…89 °C/25mmHg
Density: 1,097 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00008113
CAS Number: 100-82-3
EINECS: 202-891-3
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR