4-Fluorobenzylamine 98%

Supplier: Apollo Scientific
Danger

Synonyms: (4-Fluorophenyl)methanamine

PC3690-100G PC3690-25G
APOSPC3690-100GEA 43.5 EUR
APOSPC3690-100G APOSPC3690-25G
4-Fluorobenzylamine 98%
4-Fluorobenzylamine
Formula: C₇H₈FN
MW: 125,15 g/mol
Boiling Pt: 182…183 °C
Density: 1,095 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00008120
CAS Number: 140-75-0
EINECS: 205-430-4
UN: 2735
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR