Fluorocyclohexane 97%

Supplier: Apollo Scientific
Danger

PC3734-1G PC3734-250MG PC3734-5G
APOSPC3734-1GEA 0 EUR
APOSPC3734-1G APOSPC3734-250MG APOSPC3734-5G
Fluorocyclohexane 97%
Fluorocyclohexane
Formula: C₆H₁₁F
Boiling Pt: 103…105 °C
Melting Pt: 13 °C
Density: 0,928 g/cm³ (25 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00021285
CAS Number: 372-46-3
EINECS: 206-754-9
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR