N,N-Diethyl-2,3,3,3-tetrafluoropropionamide 97%

Supplier: Apollo Scientific
Warning

Synonyms: N,N-Diethyl-2,3,3,3-tetrafluoropropanamide

PC3787-25G
APOSPC3787-25GEA 82.8 EUR
APOSPC3787-25G
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide 97%
N,N-Diethyl-2,3,3,3-tetrafluoropropionamide
Formula: C₇H₁₁F₄NO
MW: 201.17 g/mol
Boiling Pt: 58 °C/5mmHg
Density: 1.228 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00069160
CAS Number: 392-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR