3-Fluoro-4-methoxybenzoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC3817EF-100G
PC3817EF-25G
PC3817EF-5G
APOSPC3817EF-100GEA
40.8
EUR
APOSPC3817EF-100G
APOSPC3817EF-25G
APOSPC3817EF-5G
3-Fluoro-4-methoxybenzoic acid 98%
3-Fluoro-4-methoxybenzoic acid
Formula:
C₈H₇FO₃ MW: 170,14 g/mol Melting Pt: 209…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060675 CAS Number: 403-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...