4-Fluoro-o-toluidine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Amino-5-fluorotoluene, 4-Fluoro-2-methylaniline
PC3819-100G
PC3819-250G
PC3819-25G
PC3819-500G
APOSPC3819-100GEA
30.6
EUR
APOSPC3819-100G
APOSPC3819-250G
APOSPC3819-25G
APOSPC3819-500G
4-Fluoro-o-toluidine 97%
4-Fluoro-o-toluidine
Formula:
C₇H₈FN MW: 125,15 g/mol Boiling Pt: 90…92 °C/16mmHg Density: 1,126 g/cm³ (25 °C) Flash Pt: 87 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007832 CAS Number: 452-71-1 EINECS: 207-208-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...