3-Fluoro-p-toluonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-4-methylbenzonitrile

PC3823BT-25G PC3823BT-100G
APOSPC3823BT-25GEA 14.7 EUR
APOSPC3823BT-25G APOSPC3823BT-100G
3-Fluoro-p-toluonitrile 98%
3-Fluoro-p-toluonitrile
Formula: C₈H₆FN
MW: 135,14 g/mol
Boiling Pt: 203…205 °C
Melting Pt: 47…51 °C
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00153174
CAS Number: 170572-49-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR