4-Fluoro-ɑ-methylstyrene 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Fluoro-4-(1-methylethenyl)benzene, 2-(4-Fluorophenyl)-1-propene
PC3828-10G
PC3828-1G
PC3828-25G
APOSPC3828-10GEA
48
EUR
APOSPC3828-10G
APOSPC3828-1G
APOSPC3828-25G
4-Fluoro-ɑ-methylstyrene 97%
4-Fluoro-alpha-methylstyrene
Formula:
C₉H₉F MW: 136,17 g/mol Density: 1,03 g/cm³ (25 °C) Flash Pt: 53 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042297 CAS Number: 350-40-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...