1-Fluoronaphthalene 99%

Supplier: Apollo Scientific
Warning

PC3830-100G PC3830-1KG PC3830-500G
APOSPC3830-100GEA 26.1 EUR
APOSPC3830-100G APOSPC3830-1KG APOSPC3830-500G
1-Fluoronaphthalene 99%
1-Fluoronaphthalene
Formula: C₁₀H₇F
Boiling Pt: 215…217 °C
Melting Pt: –9 °C
Density: 1,332 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00003873
CAS Number: 321-38-0
EINECS: 206-287-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR