2H-Perfluoro-5,8,11,14,17-pentamethyl-3,6,9,12,15,18-hexaoxahenicosane 95%

Supplier: Apollo Scientific
Warning

PC3866-25G PC3866-5G
APOSPC3866-25GEA 579 EUR
APOSPC3866-25G APOSPC3866-5G
2H-Perfluoro-5,8,11,14,17-pentamethyl-3,6,9,12,15,18-hexaoxahenicosane 95%
2H-Perfluoro-5,8,11,14,17-pentamethyl-3,6,9,12,15,18-hexaoxahenicosane
Formula: C₂₀HF₄₁O₆
MW: 1116,16 g/mol
Boiling Pt: 108…110 °C/1mmHg
Melting Pt: –72 °C
Density: 1,796 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD06248658
CAS Number: 55154-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR