Ethyl-3-bromo-4-fluorobenzoate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 3-bromo-4-fluorobenzoate

PC3872-1G PC3872-5G
APOSPC3872-1GEA 66.9 EUR
APOSPC3872-1G APOSPC3872-5G
Ethyl-3-bromo-4-fluorobenzoate
Ethyl 3-bromo-4-fluorobenzoate
Formula: C₉H₈BrFO₂
MW: 247,06 g/mol
Melting Pt: 44…46 °C
Storage Temperature: Ambient
MDL Number: MFCD06204378
CAS Number: 23233-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR