Potassium perfluorobutane-1-sulphonate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Nonafluoro-1-butanesulphonic acid potassium salt, Potassium Perfluoro-1-butanesulphonate, Potassium Nonafluoro-1-butanesulphonate, Perfluoro-1-butanesulphonic acid potassium salt, Potassium nonafluoro-1-butanesulphonate
PC3874-100G
PC3874-500G
APOSPC3874-100GEA
82.9
EUR
APOSPC3874-100G
APOSPC3874-500G
Potassium perfluorobutane-1-sulphonate
Potassium perfluorobutane-1-sulphonate
Additive for polymers. Soluble in water, acetone, methanol & ethyl acetate
Formula:
C₄F₉KO₃S MW: 338,1899 g/mol Melting Pt: 300 °C (dec.) Density: 0,69 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007515 CAS Number: 29420-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...