5-Fluoro-2-nitrobenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC3885D-100G
PC3885D-250G
PC3885D-25G
PC3885D-500G
APOSPC3885D-100GEA
108
EUR
APOSPC3885D-100G
APOSPC3885D-250G
APOSPC3885D-25G
APOSPC3885D-500G
5-Fluoro-2-nitrobenzaldehyde 98%
5-Fluoro-2-nitrobenzaldehyde
Formula:
C₇H₄FNO₃ Boiling Pt: 153 °C/23mmHg Melting Pt: 93…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153175 CAS Number: 395-81-3 EINECS: 206-903-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...