2,2,2-Trifluoroethyl triethylammonium triflate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,2,2-Trifluoroethyl triethylammonium trifluoromethanesulphonate
PC3902-25G
PC3902-5G
APOSPC3902-25GEA
423
EUR
APOSPC3902-25G
APOSPC3902-5G
2,2,2-Trifluoroethyl triethylammonium triflate
2,2,2-Trifluoroethyl triethylammonium triflate
Formula:
C₉H₁₇F₆NO₃S MW: 333,29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02093347 CAS Number: 380230-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...