4-Fluoro-3-nitrobenzoic acid
Supplier: Apollo Scientific
|
Warning
|
PC3935-25G
PC3935-5G
PC3935-100G
APOSPC3935-25GEA
17.6
EUR
APOSPC3935-25G
APOSPC3935-5G
APOSPC3935-100G
4-Fluoro-3-nitrobenzoic acid
4-Fluoro-3-nitrobenzoic acid
Formula:
C₇H₄FNO₄ MW: 185,11 g/mol Boiling Pt: 92 °C/15mmHg Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007058 CAS Number: 453-71-4 EINECS: 207-221-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...