6-Fluoronicotinic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-5-pyridinecarboxylic acid

PC3965-100G PC3965-25G
APOSPC3965-100GEA 437 EUR
APOSPC3965-100G APOSPC3965-25G
6-Fluoronicotinic acid 97%
6-Fluoronicotinic acid
Formula: C₆H₄FNO₂
MW: 141,1 g/mol
Melting Pt: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD01859863
CAS Number: 403-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR