2-Fluoro-4-nitrotoluene 98%

Supplier: Apollo Scientific
Danger

PC4000-1KG PC4000-250G PC4000-100G
APOSPC4000-1KGEA 0 EUR
APOSPC4000-1KG APOSPC4000-250G APOSPC4000-100G
2-Fluoro-4-nitrotoluene 98%
2-Fluoro-4-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155,13 g/mol
Boiling Pt: 65…68 °C/2mmHg
Melting Pt: 34…37 °C
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007199
CAS Number: 1427-07-2
EINECS: 215-845-2
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR