3-Bromo-5-fluoropyridin-4-amine

Supplier: Apollo Scientific

Synonyms: 4-Amino-3-bromo-5-fluoropyridine

PC400524-1G PC400524-250MG
APOSPC400524-1GEA 173 EUR
APOSPC400524-1G APOSPC400524-250MG
3-Bromo-5-fluoropyridin-4-amine
3-Bromo-5-fluoropyridin-4-amine
Formula: C5H4BrFN2
MW: 191,0000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185327
CAS Number: 1214326-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR