4-Bromo-6-(trifluoromethyl)pyrimidine

Supplier: Apollo Scientific
Warning

PC400735-5G
APOSPC400735-5GEA 179 EUR
APOSPC400735-5G
4-Bromo-6-(trifluoromethyl)pyrimidine
4-Bromo-6-(trifluoromethyl)pyrimidine
Formula: C₅H₂BrF₃N₂
MW: 226,9800 g/mol
Boiling Pt: 36…37 °C
Melting Pt: 13…15 °C
Storage Temperature: Ambient
MDL Number: MFCD10574962
CAS Number: 785777-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR