2-Amino-3,5-difluorobenzoic acid
Supplier: Apollo Scientific
|
Warning
|
PC400736-25G
PC400736-5G
APOSPC400736-25GEA
321
EUR
APOSPC400736-25G
APOSPC400736-5G
2-Amino-3,5-difluorobenzoic acid
2-Amino-3,5-difluorobenzoic acid
Formula:
C₇H₅F₂NO₂ MW: 173,1200 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01569541 CAS Number: 126674-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...