5-Nitro-2-trifluoromethyl-1H-benzoimidazole
Supplier: Apollo Scientific
|
Danger
|
PC401027-1G
PC401027-5G
APOSPC401027-1GEA
51
EUR
APOSPC401027-1G
APOSPC401027-5G
5-Nitro-2-trifluoromethyl-1H-benzoimidazole
5-Nitro-2-trifluoromethyl-1H-benzoimidazole
Formula:
C₈H₄F₃N₃O₂ MW: 231,14 g/mol Melting Pt: 154…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00458655 CAS Number: 327-19-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...