4-Bromo-7-fluoro-1H-indazole 96+%

Supplier: Apollo Scientific
Warning

PC402010-1G PC402010-250MG
APOSPC402010-1GEA 510 EUR
APOSPC402010-1G APOSPC402010-250MG
4-Bromo-7-fluoro-1H-indazole 96+%
4-Bromo-7-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
MW: 215,0225 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658655
CAS Number: 1186334-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR