6-Fluoroquinolin-4-ylamine

Supplier: Apollo Scientific

Synonyms: 4-Amino-6-fluoroquinoline

PC404560-1G PC404560-5G
APOSPC404560-1GEA 344 EUR
APOSPC404560-1G APOSPC404560-5G
6-Fluoroquinolin-4-ylamine
6-Fluoroquinolin-4-ylamine
Formula: C₉H₇FN₂
MW: 162,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664759
CAS Number: 874800-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR