3-Fluoropyrrolidine hydrochloride

Supplier: Apollo Scientific
Warning

PC405007-1G PC405007-5G
APOSPC405007-1GEA 24.8 EUR
APOSPC405007-1G APOSPC405007-5G
3-Fluoropyrrolidine hydrochloride
3-Fluoropyrrolidine hydrochloride
Formula: C4H9ClFN
MW: 125.5724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038329
CAS Number: 169750-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR