3-Chloro-5-(pentafluorothio)benzonitrile 97%
Supplier: Apollo Scientific
PC405668-1G
PC405668-5G
APOSPC405668-1GEA
348
EUR
APOSPC405668-1G
APOSPC405668-5G
3-Chloro-5-(pentafluorothio)benzonitrile 97%
3-Chloro-5-(pentafluorothio)benzonitrile
Formula:
C₇H₃ClF₅NS MW: 263,62 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22201047 CAS Number: 159727-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...