2-Fluorophenol 99%

Supplier: Apollo Scientific
Warning

PC4070-100G PC4070-500G PC4070-1KG
APOSPC4070-100GEA 50.9 EUR
APOSPC4070-100G APOSPC4070-500G APOSPC4070-1KG
2-Fluorophenol 99%
2-Fluorophenol
Formula: C₆H₅FO
Boiling Pt: 171…172 °C
Melting Pt: 14…16 °C
Density: 1,256 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00002155
CAS Number: 367-12-4
EINECS: 206-681-2
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR