3-(Trifluoroacetyl)indole
Supplier: Apollo Scientific
PC408009-10G
PC408009-1G
PC408009-5G
APOSPC408009-10GEA
546
EUR
APOSPC408009-10G
APOSPC408009-1G
APOSPC408009-5G
3-(Trifluoroacetyl)indole
3-(Trifluoroacetyl)indole
Formula:
C₁₀H₆F₃NO MW: 213.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00182114 CAS Number: 14618-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...