5-Fluoroorotic acid

Supplier: Apollo Scientific
Warning

Synonyms: 5-FOA, 2,6-Dihydroxy-5-fluoropyrimidine-4-carboxylic acid, 5-Fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid, 5-Fluorouracil-4-carboxylic acid

PC408010-25G PC408010-5G PC408010-1G
APOSPC408010-25GEA 0 EUR
APOSPC408010-25G APOSPC408010-5G APOSPC408010-1G
5-Fluoroorotic acid
5-Fluoroorotic acid
Formula: C₅H₃FN₂O₄
MW: 174,09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00042526
CAS Number: 703-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR