5,5,6,6,6-Pentafluorohexan-1-ol 95%

Supplier: Apollo Scientific
Warning

PC408150-1G PC408150-5G PC408150-250MG
APOSPC408150-1GEA 82.9 EUR
APOSPC408150-1G APOSPC408150-5G APOSPC408150-250MG
5,5,6,6,6-Pentafluorohexan-1-ol 95%
5,5,6,6,6-Pentafluorohexan-1-ol
Formula: C₆H₉F₅O
Storage Temperature: Ambient
MDL Number: MFCD06248629
CAS Number: 58556-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR