2-Fluoro-6-phenoxybenzonitrile 95%
Supplier: Apollo Scientific
|
Warning
|
PC4103-25G
PC4103-5G
PC4103-100G
APOSPC4103-25GEA
63.8
EUR
APOSPC4103-25G
APOSPC4103-5G
APOSPC4103-100G
2-Fluoro-6-phenoxybenzonitrile 95%
2-Fluoro-6-phenoxybenzonitrile
Formula:
C₁₃H₈FNO Melting Pt: 90…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068201 CAS Number: 175204-06-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...